Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methyl-5-pyridineboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (6-Methylpyridin-3-yl)boronic acid

H66327-06 H66327-03 H66327-MD
AAH66327-MDEA 46.32 USD InStock
AAH66327-MD AAH66327-06 AAH66327-03
2-Methyl-5-pyridineboronic acid 97%
2-Methyl-5-pyridineboronic acid


Formula: C₆H₈BNO₂
MW: 136.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06801736
CAS Number: 659742-21-9
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty