Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-N-fmoc-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Fmoc-Phe(2-Br)-OH

H66424-14 H66424-03 H66424-06
AAH66424-14EA 375.99 USD InStock
AAH66424-14 AAH66424-03 AAH66424-06
2-Bromo-N-fmoc-L-phenylalanine 95%
2-Bromo-N-fmoc-L-phenylalanine


Formula: C₂₄H₂₀BrNO₄
MW: 466.331 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01311743
CAS Number: 220497-47-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty