Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitroindole-2-carboxylic acid 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Nitro-1H-indole-2-carboxylic acid

H66441-06 H66441-03
AAH66441-03EA 60.89 USD InStock
AAH66441-03 AAH66441-06
5-Nitroindole-2-carboxylic acid 96%
5-Nitroindole-2-carboxylic acid


Formula: C₉H₆N₂O₄
MW: 206.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236372
CAS Number: 16730-20-4
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty