Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aminophthalide 97%

Supplier: Thermo Scientific Chemicals
Warning

H66457-03 H66457-06
AAH66457-06EA 903.99 USD InStock
AAH66457-06 AAH66457-03
5-Aminophthalide 97%
5-Aminophthalide


Formula: C₈H₇NO₂
MW: 149.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00778315
CAS Number: 65399-05-5
EINECS: 265-731-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty