Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Y5, Organic Photovoltaics (OPV) reagent

Supplier: Thermo Scientific Chemicals

H66460-ME H66460-MD H66460-MC
AAH66460-MCEA 621.99 USD InStock
AAH66460-MC AAH66460-ME AAH66460-MD
Y5, Organic Photovoltaics (OPV) reagent
Y5


Formula: C₈₂H₉₀N₈O₂S₅
MW: 1379.97 g/mol
CAS Number: 2304444-48-0
Order Now
Specification Test Results
Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1.41 ± 0.02 eV

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty