Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-ɑ-Boc-L-tryptophanol 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-alpha-(tert-Butoxycarbonyl)-L-tryptophanol(S)-2-(tert-Butoxycarbonyl)amino-3-(3-indolyl)propanol(S)-2-(Boc-amino)-3-(3-indolyl)propanol

H66571-03 H66571-06 H66571-MD
AAH66571-MDEA 30.39 USD InStock
AAH66571-MD AAH66571-06 AAH66571-03
N-ɑ-Boc-L-tryptophanol 95%
N-alpha-Boc-L-tryptophanol


Formula: C₁₆H₂₂N₂O₃
MW: 290.36 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00235953
CAS Number: 82689-19-8
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty