Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-D-aspartic acid-1-methyl ester 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Boc-D-Asp-OMe(R)-3-(Boc-amino)-4-methoxy-4-oxobutyric acid

H66573-14 H66573-06
AAH66573-14EA 733.99 USD InStock
AAH66573-14 AAH66573-06
N-Boc-D-aspartic acid-1-methyl ester 97%
N-Boc-D-aspartic acid-1-methyl ester


Formula: C₁₀H₁₇NO₆
MW: 247.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094776
CAS Number: 137130-65-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty