Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-Amino-3-cyclopentylpropanoic acid 95%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Cpa-OH3-Cyclopentyl-L-alanine

H66600-03 H66600-MD
AAH66600-MDEA 109.99 USD InStock
AAH66600-MD AAH66600-03
(S)-2-Amino-3-cyclopentylpropanoic acid 95%
(S)-2-Amino-3-cyclopentylpropanoic acid


Formula: C8H15NO2
MW: 157.21 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01632069
CAS Number: 99295-82-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty