Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Piperazin-1-yl)-1H-indole 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(1-Piperazinyl)indole

H66602-03 H66602-MD
AAH66602-MDEA 124.99 USD InStock
AAH66602-MD AAH66602-03
4-(Piperazin-1-yl)-1H-indole 95%
4-(Piperazin-1-yl)-1H-indole


Formula: C₁₂H₁₅N₃
MW: 201.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00831776
CAS Number: 84807-09-0
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty