Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Valine ethyl ester hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Synonyms: H-D-Val-OEt·HCl(R)-Ethyl 2-amino-3-methylbutanoate hydrochloride

H66619-06 H66619-14 H66619-03
AAH66619-14EA 477.99 USD InStock
AAH66619-14 AAH66619-03 AAH66619-06
D-Valine ethyl ester hydrochloride 97%
D-Valine ethyl ester hydrochloride


Formula: C₇H₁₆ClNO₂
MW: 181.66 g/mol
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00191043
CAS Number: 73913-64-1
EINECS: 277-635-7
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty