Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(2-Methyl-6-nitrophenyl)acetic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H66654-MD H66654-03
AAH66654-MDEA 52.19 USD InStock
AAH66654-MD AAH66654-03
2-(2-Methyl-6-nitrophenyl)acetic acid 95%
2-(2-Methyl-6-nitrophenyl)acetic acid


Formula: C₉H₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00066313
CAS Number: 23876-18-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty