Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-IDTBr, Organic Photovoltaics (OPV) reagent

Supplier: Thermo Scientific Chemicals

H66666-ME H66666-MC H66666-MD
AAH66666-MDEA 1125.99 USD InStock
AAH66666-MD AAH66666-ME AAH66666-MC
o-IDTBr, Organic Photovoltaics (OPV) reagent
o-IDTBr


Formula: C₇₂H₈₈N₆O₂S₈
MW: 1326.03 g/mol
CAS Number: 2077945-91-4
Order Now
Specification Test Results
Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1.60 ± 0.02 eV

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty