Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Benzyloxycarbonyl-3-bromopyrrolidine 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

H66676-MD H66676-06 H66676-03
AAH66676-06EA 652.99 USD InStock
AAH66676-06 AAH66676-MD AAH66676-03
1-Benzyloxycarbonyl-3-bromopyrrolidine 90%, Technical Grade
1-Benzyloxycarbonyl-3-bromopyrrolidine


Formula: C₁₂H₁₄BrNO₂
MW: 284.153 g/mol
Density: 1.395
Flash Pt: 110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD13181593
CAS Number: 220212-12-4
Order Now
Specification Test Results
Assay Minimum 90

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty