Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-S-trityl-L-cysteine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Ac-Cys(Trt)-OH

H66689-06 H66689-03
AAH66689-03EA 116.24 USD InStock
AAH66689-03 AAH66689-06
N-Acetyl-S-trityl-L-cysteine 95%
N-Acetyl-S-trityl-L-cysteine


Formula: C₂₄H₂₃NO₃S
MW: 405.512 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00236747
CAS Number: 27486-87-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty