Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-D-asp-Otbu 98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Boc-D-aspartic acid 1-tert-butyl ester(R)-4-(tert-Butoxy)-3-((tert-butoxycarbonyl)amino)-4-oxobutanoic acid

H66716-MD H66716-06 H66716-03
AAH66716-03EA 67.39 USD InStock
AAH66716-03 AAH66716-06 AAH66716-MD
Boc-D-asp-Otbu 98%
Boc-D-asp-Otbu


Formula: C₁₃H₂₃NO₆
MW: 289.328 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00236827
CAS Number: 77004-75-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty