Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Z-Tyr(tBu)-OH

H66762-14 H66762-22 H66762-06
AAH66762-22EA 516.99 USD InStock
AAH66762-22 AAH66762-14 AAH66762-06
N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine 98%
N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine


Formula: C₂₁H₂₅NO₅
MW: 371.433 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065701
CAS Number: 5545-54-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty