Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-isoleucine 98% (by HPLC)

Supplier: Thermo Scientific Chemicals
Synonyms: Ac-Ile-OH

H66771-22 H66771-06 H66771-14
AAH66771-06EA 79.49 USD InStock
AAH66771-06 AAH66771-14 AAH66771-22
N-Acetyl-L-isoleucine 98% (by HPLC)
N-Acetyl-L-isoleucine


Formula: C₈H₁₅NO₃
MW: 173.212 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00066072
CAS Number: 3077-46-1
EINECS: 221-364-9
Order Now
Specification Test Results
Assay (HPLC) 97.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty