Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propyl benzoate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Benzoic acid propyl ester

H66793-30 H66793-18
AAH66793-18EA 40.43 USD InStock
AAH66793-18 AAH66793-30
Propyl benzoate 99%
Propyl benzoate


Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 231 °C
Density: 1.026
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009370
CAS Number: 2315-68-6
EINECS: 219-020-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty