Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Amino-4-chloro-7H-pyrrolo[2,3-d]pyrimidine

H66806-MD H66806-03
AAH66806-03EA 221.99 USD InStock
AAH66806-03 AAH66806-MD
4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine 97%
4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-2-amine


Formula: C6H5ClN4
MW: 168.58 g/mol
Melting Pt: 215…220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07369229
CAS Number: 84955-31-7
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty