Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)iminodiacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-Boc-iminodiacetic Acid

H66868-14 H66868-06
AAH66868-14EA 272.99 USD InStock
AAH66868-14 AAH66868-06
N-(tert-Butoxycarbonyl)iminodiacetic acid 98%
N-(tert-Butoxycarbonyl)iminodiacetic acid


Formula: C₉H₁₅NO₆
MW: 233.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00800351
CAS Number: 56074-20-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty