Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-trans-4-(fmoc-amino)-L-proline 97%

Supplier: Thermo Scientific Chemicals
Synonyms: (2S,4R)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acidN-BOC-trans-4-N-FMOC-amino-L-proline

H66878-MD H66878-03
AAH66878-MDEA 92.89 USD InStock
AAH66878-MD AAH66878-03
N-Boc-trans-4-(fmoc-amino)-L-proline 97%
N-Boc-trans-4-(fmoc-amino)-L-proline


Formula: C₂₅H₂₈N₂O₆
MW: 452.507 g/mol
Melting Pt: 197…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00673783
CAS Number: 176486-63-8
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty