Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Tosyl-l-proline 94%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tos-Pro-OH(2S)-1-[(4-methylphenyl)sulfonyl]pyrrolidine-2-carboxylic acid

H66902-14 H66902-06
AAH66902-06EA 75.56 USD InStock
AAH66902-06 AAH66902-14
N-Tosyl-l-proline 94%
N-Tosyl-l-proline


Formula: C₁₂H₁₅NO₄S
MW: 269.315 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00238392
CAS Number: 51077-01-1
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty