Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-N-methyl-1,3-diaminopropane 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-Methylpropane-1,3-diamine, N-BOC protected3-(N-Boc-N-methylamino)propylamine

H66932-03 H66932-MD
AAH66932-03EA 149.99 USD InStock
AAH66932-03 AAH66932-MD
N-Boc-N-methyl-1,3-diaminopropane 95%
N-Boc-N-methyl-1,3-diaminopropane


Formula: C₉H₂₀N₂O₂
MW: 188.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631189
CAS Number: 150349-36-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty