Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Tryptophan ethyl ester hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-2-Amino-3-(3-indolyl)propionic Acid Ethyl Ester HydrochlorideH-Trp-OEt.HCl

H66953-22 H66953-14 H66953-36
AAH66953-22EA 142.99 USD InStock
AAH66953-22 AAH66953-14 AAH66953-36
L-Tryptophan ethyl ester hydrochloride 97%
L-Tryptophan ethyl ester hydrochloride


Formula: C₁₃H₁₆N₂O₂
MW: 268.74 g/mol
Melting Pt: 220…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00038993
CAS Number: 2899-28-7
EINECS: 220-786-0
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty