Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Methylguanidine xhydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methylguanidine hydrochloride

J60033-06 J60033-14 J60033-14 J60033-06
AAJ60033-14EA 248.55 USD InStock
CAAAJ60033-06 CAAAJ60033-14 AAJ60033-14 AAJ60033-06
1-Methylguanidine xhydrochloride 98%
1-Methylguanidine xhydrochloride
Crystalline Powder


Formula: C₂H₈ClN₃
MW: 109.557 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012576
CAS Number: 22661-87-6
EINECS: 245-147-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty