Synonyms:
2-IT
, 2-IT HCl
, 2-Iminothiolane hydrochloride
, 2-Thiolanimine hydrochloride
J60131-MD
J60131-03
J60131-MD
J60131-03
AAJ60131-MDEA
187.71
USD
InStock
AAJ60131-MD
CAAAJ60131-03
AAJ60131-03
CAAAJ60131-MD
2-Iminothiolane hydrochloride
2-Iminothiolane hydrochloride (Traut's Reagent)
Protein modification and cross-linking reagent, cleavable by thiols
Powder