Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Yeast Extract, Thermo Scientific

Supplier: Thermo Scientific
J60287-A1 J60287-A1
AAJ60287-A1EA 261.44 USD InStock
CAAAJ60287-A1 AAJ60287-A1
Yeast Extract, Thermo Scientific
Media for Microbiology Dehydrated Media for Microbiology
Powder

CAS No.: 8013-01-2
Physical Form: Powder
Appearance: Yellow to brown
MDL No.: MFCD00132599

Microbial media

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty