Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium mercaptoacetate (sodium thioglycolate) 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Thioglycolic AcidMercaptoacetic acid, sodium saltThioglycolic Acid Sodium Salt

J60290-36 J60290-14 J60290-A1 J60290-22 J60290-14 J60290-22 J60290-A1
10813-634EA 265.73 USD InStock
CAAAJ60290-A1 CAAAJ60290-22 10813-634 CAAAJ60290-14 AAJ60290-22 AAJ60290-A1 AAJ60290-14
Sodium mercaptoacetate (sodium thioglycolate) 98%
Sodium mercaptoacetate (Sodium thioglycolate)
Reagent that protects tryptophan in amino acid analysis

Powder


Formula: HSCH₂COONa
MW: 114.1 g/mol
Melting Pt: >300 °C
Storage Temperature: Freezer
MDL Number: MFCD00043386
CAS Number: 367-51-1
EINECS: 206-696-4
UN: 2923
ADR: "8",III
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty