Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adenosine-5′-triphosphate disodium salt hydrate 98%, ultrapure

Supplier: Thermo Scientific Chemicals

J60336-14 J60336-14 J60336-22 J60336-22
AAJ60336-22EA 971.68 USD InStock
CAAAJ60336-22 AAJ60336-22 AAJ60336-14 CAAAJ60336-14
Adenosine-5′-triphosphate disodium salt hydrate 98%, ultrapure
Adenosine-5'-triphosphate disodium salt hydrate
Powder


Formula: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MW: 551.2 g/mol (anhy)
Storage Temperature: Freezer
CAS Number: 51963-61-2
EINECS: 213-579-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty