Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chondroitin sulfate ≥90%

Supplier: Thermo Scientific Chemicals

J60341-06 J60341-06 J60341-03 J60341-14 J60341-03 J60341-14
AAJ60341-03EA 42.67 USD InStock
AAJ60341-03 AAJ60341-14 AAJ60341-06 CAAAJ60341-06 CAAAJ60341-03 CAAAJ60341-14
Chondroitin sulfate ≥90%
Chondroitin sulfate
A mucopolysaccharide with N-acetylchondrosine as a repeating unit and with one sulfate group per disaccharide unit; acts as the flexible connecting matrix between the tough protein filaments in cartilage to form a polymeric system.A naturocetic agent

Powder


Storage Temperature: Ambient
MDL Number: MFCD00146419
CAS Number: 9007-28-7
EINECS: 232-696-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty