Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Phenylenediamine dihydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,2-Diaminobenzene dihydrochloride 1,2-Phenylenediamine dihydrochloride 1,2-Phenylenediammonium dichloride Benzene-1,2-diamine dihydrochloride

J60354-22 J60354-14 J60354-22 J60354-14
AAJ60354-14EA 104.85 USD InStock
AAJ60354-14 CAAAJ60354-22 AAJ60354-22 CAAAJ60354-14
o-Phenylenediamine dihydrochloride 98+%
o-Phenylenediamine dihydrochloride
A peroxidase substrate for ELISA procedures

Powder


Formula: C₆H₁₀Cl₂N₂
MW: 181.06 g/mol
Melting Pt: 205 °C
Storage Temperature: Freezer
MDL Number: MFCD00012966
CAS Number: 615-28-1
EINECS: 210-418-7
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty