Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate 97%

Supplier: Thermo Scientific Chemicals
Synonyms: β-NADPH tetrasodium salt tetrahydrate

J60387-MC J60387-ME J60387-ME J60387-MC
AAJ60387-MEEA 785.16 USD InStock
CAAAJ60387-MC AAJ60387-ME AAJ60387-MC CAAAJ60387-ME
β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate 97%
β-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate
Applications: beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.

  • Store in cool place
  • Keep container tightly closed in a dry and well-ventilated place
  • Light sensitive
  • Hygroscopic

Purity: 98+%
Molecular Formula: C21H26N7Na4O17P3.xH2O
Formula Weight: 905.42 (833.35anhy)
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD07366294


Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty