Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-D-(+)-Melibiose

Supplier: Thermo Scientific Chemicals
Synonyms: 6-O-alpha-D-Galactopyranosyl-D-glucose

J60439-06 J60439-06
AAJ60439-06EA 234.36 USD InStock
AAJ60439-06 CAAAJ60439-06
α-D-(+)-Melibiose
alpha-D-(+)-Melibiose
Appearance: White crystalline powder
Solubility: Soluble in water at 100mg/ml


Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00198188
CAS Number: 585-99-9
EINECS: 209-568-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty