Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use, precipitating

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: TMB

J60461-AP J60461-AU J60461-AP J60461-AU
AAJ60461-AUEA 385.52 USD InStock
CAAAJ60461-AU AAJ60461-AU AAJ60461-AP CAAAJ60461-AP
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use, precipitating
3,3',5,5'-Tetramethylbenzidine (TMB)
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result

Liquid


Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00007748
CAS Number: 54827-17-7
EINECS: 259-364-6
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty