Would you like to log in again?
LB Agar Plate with Kanamycin, Thermo Scientific
Supplier: Thermo Scientific
J60487-EQF J60540-EQF J60540-EQF J60487-EQF
AAJ60487-EQFEA 186.04 USD InStock
AAJ60487-EQF CAAAJ60487-EQF AAJ60540-EQF CAAAJ60540-EQF
LB Agar Plate with Kanamycin, Thermo Scientific
Media for Microbiology Prepared Media for Microbiology
This LB Agar plate with kanamycin is suitable for selective cultivation of <italic>E.coli </italic>strains that contain plasmids conferring kanamycin resistance.
  • Solid
  • Not miscible or difficult to mix in water
The bacteria can be used for DNA plasmid production or production of recombinant proteins.
Caution: Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty