Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hyaluronic acid (from bovine vitreous humor)

Supplier: Thermo Scientific Chemicals

J60566-MC J60566-MA J60566-MA J60566-MF J60566-MC J60566-MF
AAJ60566-MFEA 612.79 USD InStock
CAAAJ60566-MA CAAAJ60566-MF CAAAJ60566-MC AAJ60566-MF AAJ60566-MA AAJ60566-MC
Hyaluronic acid (from bovine vitreous humor)
Hyaluronic acid
A natural high-viscosity polymer of acetylglucosamine and glucuronic acid

Lyophilized powder


Formula: C₂₈H₄₄N₂O₂₃
Storage Temperature: Refrigerator
MDL Number: MFCD00131348
CAS Number: 9004-61-9
EINECS: 232-678-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty