Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Kanamycin sulfate cell culture reagent

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Kanamycin MonosulphateKanamycin AKanamycin Monosulfate

J60668-06 J60668-09 J60668-06 J60668-03 J60668-03 J60668-09
AAJ60668-09EA 269.75 USD InStock
AAJ60668-09 CAAAJ60668-03 CAAAJ60668-06 CAAAJ60668-09 AAJ60668-06 AAJ60668-03
Kanamycin sulfate cell culture reagent
Kanamycin sulphate
Powder


Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070253
CAS Number: 25389-94-0
EINECS: 246-933-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty