Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adenosine-5'-diphosphate monopotassium salt dihydrate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Potassium (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-4-hydroxy-5-(((hydroxy(phosphonooxy)phosphoryl)oxy)methyl)tetrahydrofuran-3-olate dihydrateADP-K5'-ADP-K

J60672-03 J60672-06 J60672-03 J60672-06
AAJ60672-06EA 310.32 USD InStock
CAAAJ60672-03 CAAAJ60672-06 AAJ60672-06 AAJ60672-03
Adenosine-5'-diphosphate monopotassium salt dihydrate 99%
Adenosine-5'-diphosphate monopotassium salt dihydrate
Crystalline powder


Formula: C₁₀H₁₄KN₅O₁₀P₂.2H₂O
MW: 501.33 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00066472
CAS Number: 72696-48-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty