Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Thiouridine ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-thioxo-3,4-dihydropyrimidin-2(1H)-one

J60679-MC J60679-MD J60679-MC
AAJ60679-MDEA 789.49 USD InStock
AAJ60679-MD CAAAJ60679-MC AAJ60679-MC
4-Thiouridine ≥98%
4-Thiouridine
Crystalline


Formula: C₉H₁₂N₂O₅S
MW: 260.264 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00006538
CAS Number: 13957-31-8
EINECS: 237-735-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty