Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium acetate 5 M in aqueous solution for molecular biology

Supplier: Thermo Scientific Chemicals
Synonyms: Acetic acid ammonium salt

J60688-AE
AAJ60688-AEEA 124.8 USD InStock
AAJ60688-AE
Ammonium acetate 5 M in aqueous solution for molecular biology
Ammonium acetate
Ammonium acetate is used for routine precipitation of nucleic acids, and is especially useful for reducing the coprecipitation of unwanted dNTPs and contaminating oligosaccharides.

It is used as an important reagent for studying molecular biology, biological buffers, reagents and DNA and RNA purification. It is a popular buffer for mobile phases for HPLC with ESLD detection, for ESI mass spectrometry of proteins and other molecules, and has been used to replace cell buffers with non-volatile salts. Ammonium acetate is also used in protein studies and protein preparation. It can be used in the protein purification steps of dialysis to remove contaminants through diffusion and, when combined with distilled water, as a protein precipitating agent. In organic chemistry, ammonium acetate solution is useful as a catalyst in the knoevenagel condensation and as a source of ammonia in the borch reaction. Additionally, it is occasionally used commercially as a biodegradable de-icing agent and as an additive in food as an acidity regulator.


Formula: C₂H₇NO₂
MW: 77.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00013066
CAS Number: 631-61-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/04/16640604.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty