Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Glutamine 99+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-(-)-Glutamine H-D-Gln-OH

J60784-03 J60784-03 J60784-14 J60784-06 J60784-14 J60784-06
AAJ60784-03EA 38.24 USD InStock
AAJ60784-03 AAJ60784-14 CAAAJ60784-06 CAAAJ60784-03 CAAAJ60784-14 AAJ60784-06
D-(-)-Glutamine 99+%
D(-)-Glutamine
Kinase inhibitor; has antivasospastic properties

Powder


Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065607
CAS Number: 5959-95-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty