Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Glucagon (1-29)

Supplier: Thermo Scientific
J60827-EXD J60827-EXD J60827-MCR J60827-MCR
AAJ60827-MCREA 320.59 USD InStock
CAAAJ60827-EXD AAJ60827-MCR AAJ60827-EXD CAAAJ60827-MCR
Human Glucagon (1-29)
Proteins and Peptides
CAS No.: 16941-32-5
Molecular Formula: C153H225N43O49S
Formula Weight: 3482.74
Physical Form: Powder
Appearance: White

A peptide hormone that plays a role in maintaining glucose homeostasis

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty