Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Folic acid, crystalline

Supplier: Thermo Scientific Chemicals
Synonyms: Pteroyl-L-glutamic acidVitamin B9PteGluVitamin M(S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid

J60833-14 J60833-22 J60833-14 J60833-22
AAJ60833-14EA 120.73 USD InStock
AAJ60833-14 AAJ60833-22 CAAAJ60833-14 CAAAJ60833-22
Folic acid, crystalline
Folic acid
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells

Crystalline


Formula: C₁₉H₁₉N₇O₆
MW: 441.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079305
CAS Number: 59-30-3
EINECS: 200-419-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty