Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DNA (Deoxyribonucleic acid) (from salmon testes)

Supplier: Thermo Scientific Chemicals

J60840-03 J60840-03
AAJ60840-03EA 370.18 USD InStock
CAAAJ60840-03 AAJ60840-03
DNA (Deoxyribonucleic acid) (from salmon testes)
DNA (Deoxyribonucleic acid)
Lyophilized powder


Storage Temperature: Ambient
MDL Number: MFCD00283430
CAS Number: 68938-01-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty