Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Clofarabine 99+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Chloro-9-(2-deoxy-2-fluoro-beta-D-arabinofuranosyl)adenine

J60927-MD J60927-MD
AAJ60927-MDEA 182.61 USD InStock
CAAAJ60927-MD AAJ60927-MD
Clofarabine 99+%
Clofarabine
A purine nucleoside anti-metabolite. Toxic to nondividing lymphocytes and monocytes

Powder


Formula: C₁₀H₁₁ClFN₅O₃
MW: 303.68 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00871077
CAS Number: 123318-82-1
UN: 2811
ADR: "6.1",III
Order Now
Specification Test Results
Assay Minimum 99

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty