Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adenosine-3',5'-cyclic monophosphate (cAMP) 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Adenosine 3′,5′-cyclophosphate3',5'-cAMP3′,5′-Cyclic AMP(4aR,6R,7R,7aS)-6-(6-Amino-9H-purin-9-yl)-2,7-dihydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinine 2-oxide

J60936-06 J60936-06 J60936-03 J60936-03
AAJ60936-03EA 215.81 USD InStock
CAAAJ60936-03 CAAAJ60936-06 AAJ60936-03 AAJ60936-06
Adenosine-3',5'-cyclic monophosphate (cAMP) 98%
Adenosine-3',5'-cyclic monophosphate (cAMP)
Crystalline powder


Formula: C₁₀H₁₂N₅O₆P
MW: 329.21 g/mol
Melting Pt: 260 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00005845
CAS Number: 60-92-4
EINECS: 200-492-9
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty