Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-[(S)-1-Hydroxyethyl]pyridine

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-1-(Pyridin-2-yl)ethanol(S)-1-(2-Pyridyl)ethanol(S)-alpha-Methyl-2-pyridinemethanol

J60980-03 J60980-03
AAJ60980-03EA 159.27 USD InStock
CAAAJ60980-03 AAJ60980-03
2-[(S)-1-Hydroxyethyl]pyridine
2-[(S)-1-Hydroxyethyl]pyridine
For synthesis of optically active products

Solid


Formula: C₇H₉NO
MW: 123.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06795465
CAS Number: 59042-90-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty