Synonyms:
Tetrazole
J61034-AO
J61034-AH
J61034-AO
J61034-AH
AAJ61034-AOEA
631.61
USD
InStock
AAJ61034-AO
CAAAJ61034-AO
AAJ61034-AH
CAAAJ61034-AH
1H-Tetrazole 0.45 mol/l in acetonitrile
1H-Tetrazole
Coupling reagent for preparation of polynucleotides
Liquid