Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1H-Tetrazole 0.45 mol/l in acetonitrile

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tetrazole

J61034-AO J61034-AH J61034-AO J61034-AH
AAJ61034-AOEA 631.61 USD InStock
AAJ61034-AO CAAAJ61034-AO AAJ61034-AH CAAAJ61034-AH
1H-Tetrazole 0.45 mol/l in acetonitrile
1H-Tetrazole
Coupling reagent for preparation of polynucleotides

Liquid


Formula: CH₂N₄
MW: 70.05 g/mol
Density: 0.798
Storage Temperature: Ambient
MDL Number: MFCD00005247
CAS Number: 288-94-8
EINECS: 206-023-4
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty