Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thioflavin T ≥75%, tech.

Supplier: Thermo Scientific Chemicals
Synonyms: Setoflavine TThioflavine T

J61043-06 J61043-14 J61043-06 J61043-14
AAJ61043-06EA 26.81 USD InStock
AAJ61043-06 CAAAJ61043-06 CAAAJ61043-14 AAJ61043-14
Thioflavin T ≥75%, tech.
Thioflavin T
A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease

Powder


Formula: C₁₇H₁₉ClN₂S
MW: 318.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011944
CAS Number: 2390-54-7
EINECS: 219-228-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty